Search
Search
| Gene Symbol: Ddx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145885.1 | NP_001139357.1 | 18 - 19 | 1663 | 124 | |
| NM_010029.2 | NP_034159.1 | 17 - 18 | 1585 | 124 | ||
| GenBank mRNA | AK014844.1 | - | 17 - 18 | 1576 | 124 | |
| AK133366.1 | - | 18 - 19 | 1656 | 124 | ||
| BC137601.1 | - | 17 - 18 | 1639 | 8860620 | 124 | |
| BC144760.1 | - | 17 - 18 | 1639 | 9053308 | 124 | |
| JQ923486.1 | - | 17 - 18 | 1613 | 124 | ||
Set Membership: |
|