| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500000698.mm.1 |
NM_001253781.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000698.mm.1 |
NM_001253781.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000698.mm.1 |
NM_001253782.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000698.mm.1 |
NM_001253782.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000698.mm.1 |
NM_011169.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000698.mm.1 |
NM_011169.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000698.mm.1 |
TC1500002327.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500000683.mm.1 |
NM_011169.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000683.mm.1 |
NM_011169.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000683.mm.1 |
TC1500002327.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500000696.mm.1 |
NM_001253781.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000696.mm.1 |
NM_001253781.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000696.mm.1 |
NM_001253782.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000696.mm.1 |
NM_001253782.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000696.mm.1 |
NM_011169.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000696.mm.1 |
NM_011169.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000696.mm.1 |
TC1500002327.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500000692.mm.1 |
NM_001253781.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000692.mm.1 |
NM_001253781.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000692.mm.1 |
TC1500002327.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500000691.mm.1 |
NM_001253781.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000691.mm.1 |
NM_001253781.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000691.mm.1 |
TC1500002327.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500000694.mm.1 |
NM_001253781.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000694.mm.1 |
NM_001253781.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000694.mm.1 |
NM_001253782.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000694.mm.1 |
NM_001253782.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000694.mm.1 |
NM_011169.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000694.mm.1 |
NM_011169.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000694.mm.1 |
TC1500002327.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500000684.mm.1 |
NM_011169.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000684.mm.1 |
NM_011169.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000684.mm.1 |
TC1500002327.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500000686.mm.1 |
NM_011169.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000686.mm.1 |
NM_011169.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000686.mm.1 |
TC1500002327.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500000688.mm.1 |
NM_011169.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000688.mm.1 |
NM_011169.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000688.mm.1 |
TC1500002327.mm.1 |
MTA TC |