Search
Search

Hdac4| Gene Symbol: Hdac4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207225.1 | NP_997108.1 | 6 - 7 | 845 | 78 | |
| GenBank mRNA | AK029933.1 | - | 3 - 4 | 382 | 78 | |
| AK155250.1 | - | 7 - 8 | 1031 | 78 | ||
| BC066052.1 | - | 6 - 7 | 845 | 6827645 | 78 | |
Set Membership: |
|