| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700014496.mm.1 |
NM_001172113.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014496.mm.1 |
NM_001172113.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014496.mm.1 |
NM_001172114.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014496.mm.1 |
NM_001172114.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014496.mm.1 |
NM_153792.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014496.mm.1 |
NM_153792.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014496.mm.1 |
TC1700002797.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700014494.mm.1 |
NM_001172113.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014494.mm.1 |
NM_001172113.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014494.mm.1 |
NM_001172114.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014494.mm.1 |
NM_001172114.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014494.mm.1 |
NM_153792.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014494.mm.1 |
NM_153792.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014494.mm.1 |
TC1700002797.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700014497.mm.1 |
NM_001172113.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014497.mm.1 |
NM_001172113.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014497.mm.1 |
NM_001172114.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014497.mm.1 |
NM_001172114.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014497.mm.1 |
NM_153792.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014497.mm.1 |
NM_153792.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700014497.mm.1 |
TC1700002797.mm.1 |
MTA TC |