Search
Search

Triobp| Gene Symbol: Triobp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024716.1 | NP_001019887.1 | 4 - 5 | 370 | 60 | |
| NM_001039156.1 | NP_001034245.1 | 15 - 16 | 5137 | 60 | ||
| NM_138579.4 | NP_613045.3 | 14 - 15 | 4999 | 60 | ||
| GenBank mRNA | AK053194.1 | - | 4 - 5 | 330 | 60 | |
| AK085512.1 | - | 4 - 5 | 340 | 60 | ||
| AK155197.1 | - | 7 - 8 | 2702 | 60 | ||
| AK170390.1 | - | 4 - 5 | 316 | 60 | ||
| BC003984.1 | - | 2 - 3 | 166 | 60 | ||
| BC042760.1 | - | 4 - 5 | 309 | 60 | ||
| BC053329.1 | - | 4 - 5 | 354 | 60 | ||
| CF533172.1 | - | 4 - 5 | 442 | 60 | ||
| DQ228000.1 | - | 15 - 16 | 5137 | 60 | ||
| DQ228001.2 | - | 14 - 15 | 4999 | 60 | ||
| DQ278602.1 | - | 13 - 14 | 4618 | 60 | ||
Set Membership: |
|