Search
Search

Triobp| Gene Symbol: Triobp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024716.1 | NP_001019887.1 | 7 - 8 | 1119 | 63 | |
| NM_001039156.1 | NP_001034245.1 | 18 - 19 | 5886 | 63 | ||
| NM_138579.4 | NP_613045.3 | 17 - 18 | 5748 | 63 | ||
| GenBank mRNA | AK053194.1 | - | 7 - 8 | 1079 | 63 | |
| AK085512.1 | - | 7 - 8 | 1090 | 63 | ||
| AK129417.1 | - | 6 - 7 | 2544 | 63 | ||
| AK155197.1 | - | 10 - 11 | 3454 | 63 | ||
| AK170390.1 | - | 7 - 8 | 1065 | 63 | ||
| BC003984.1 | - | 5 - 6 | 915 | 63 | ||
| BC042760.1 | - | 7 - 8 | 1058 | 63 | ||
| BC053329.1 | - | 7 - 8 | 1103 | 63 | ||
| CA751843.1 | - | 2 - 3 | 565 | 63 | ||
| DQ228000.1 | - | 18 - 19 | 5886 | 63 | ||
| DQ228001.2 | - | 17 - 18 | 5748 | 63 | ||
| DQ278602.1 | - | 16 - 17 | 5370 | 63 | ||
Set Membership: |
|