Search
Search

Ccdc27| Gene Symbol: Ccdc27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033455.2 | NP_001028627.1 | 9 - 10 | 1553 | 97 | |
| GenBank mRNA | AK133459.1 | - | 9 - 10 | 1554 | 97 | |
| BC150930.1 | - | 9 - 10 | 1536 | 9087841 | 97 | |
| BC151167.1 | - | 9 - 10 | 1536 | 9088056 | 97 | |
Set Membership: |
|