Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01300970_g1
          See other CLPS GE Assays ›
          Gene Symbol:
          Clps
          Gene Name:
          colipase, pancreatic
          Gene Aliases:
          2200003J09Rik
          Chromosome Location:
          Chr.17: 28558214 - 28560714 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01300970_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Clps
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025469.2 NP_079745.1 2 - 3 270 71
          GenBank mRNA AF414676.1 - 2 - 3 257 71
          AF414677.1 - 2 - 3 257 71
          AK008635.1 - 2 - 3 268 71
          AK008815.1 - 2 - 3 271 71
          AK008834.1 - 2 - 3 268 71
          AK008839.1 - 2 - 3 268 71
          AK008874.1 - 2 - 3 268 71
          AK008879.1 - 2 - 3 267 71
          AK019047.1 - 2 - 3 268 71
          AK028141.1 - 2 - 3 270 71
          BC042935.1 - 2 - 3 255 6399295 71

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Clps

          Panther Classification:

          Molecular Function -

          protein-binding activity modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          digestion
          lipid catabolic process
          positive regulation of catalytic activity
          enzyme activator activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2015878 AK008834.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2015878 AK008839.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2015878 AK008874.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2015878 AK008879.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2015878 AK008635.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2015877 AK008834.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2015877 AK008839.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2015877 AK008874.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2015877 AK008879.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2015877 AF414677.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 143054 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 740158 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 22899 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179048 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2015877 AK008834.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2015877 AK008839.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2015877 AK008874.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2015877 AK008879.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2015877 AF414677.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2015878 AK008834.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2015878 AK008839.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2015878 AK008874.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2015878 AK008879.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2015878 AK008635.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P164459 AK008834.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P164459 AK008839.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P164459 AK008874.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P164459 AK008879.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P164459 AF414677.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123092 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123095 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123118 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123114 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123119 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123086 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123108 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123116 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123111 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123097 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123115 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123106 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123102 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123122 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123100 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123087 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123104 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123112 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P179048 - -
          Whole Mouse Genome G4122A A_51_P164459 AK008834.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164459 AK008839.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164459 AK008874.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164459 AK008879.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164459 AF414677.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107373 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P179048 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13061180 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13162681 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13162675 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13061179 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13064934 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13162680 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13162674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13308220 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123116 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123065 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13541548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123063 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13350931 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123111 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13162681 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123076 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123069 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123118 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123074 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123071 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13308221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13061176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13061181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13129408 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123087 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13496261 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13162675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13541544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123114 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123106 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123115 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123108 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123100 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123083 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13061179 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123121 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10449469 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449470 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449468 - -
          Affymetrix HT MG-430 PM Array Plate 1415805_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1438612_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10449467 - -
          Mouse Genome 430 2.0 Array 1438612_a_at - -
          Mouse Genome 430 2.0 Array 1438612_PM_a_at - -
          Mouse Genome 430 2.0 Array 1415805_PM_at - -
          Mouse Genome 430 2.0 Array 1415805_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 109791_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 109791_at - -
          Mouse Expression Set 430 1415805_at - -
          Mouse Expression Set 430 1438612_a_at - -
          Mouse Expression Set 430 1415805_PM_at - -
          Mouse Expression Set 430 1438612_PM_a_at - -
          Murine Genome U74v2 Set 160132_at - -
          Murine Genome U74v2 Set 160132_PM_at - -
          Mouse Genome 430A 2.0 Array 1415805_PM_at - -
          Mouse Genome 430A 2.0 Array 1415805_at - -
          Mouse Genome 430A 2.0 Array 1438612_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1438612_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1438612_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1438612_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1415805_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1415805_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1340056 - -
          Illumina mouse-6 v1.0 expression beadchip 6220524 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001340056 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006220524 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1246265 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2722659 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1246265 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2722659 - -
          mouse-6 v1.1 (Illumina) scl0001702.1_4-S - -
          mouse-6 v1.1 (Illumina) scl50115.4.1_12-S - -
          Illumina Sentrix mouse-6.v1 bead array 6220524 - -
          Illumina Sentrix mouse-6.v1 bead array 1340056 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0001702.1_4-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50115.4.1_12-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1340056 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6220524 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2722659 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1246265 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1246265 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2722659 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline