Search
Search

Chrnd| Gene Symbol: Chrnd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021600.3 | NP_067611.2 | 4 - 5 | 459 | 73 | |
| GenBank mRNA | AK081344.1 | - | 3 - 4 | 445 | 73 | |
| BC052153.1 | - | 4 - 5 | 404 | 6336578 | 73 | |
| BC141376.1 | - | 4 - 5 | 387 | 9055989 | 73 | |
| BC141387.1 | - | 4 - 5 | 387 | 9056000 | 73 | |
| K02582.1 | - | 4 - 5 | 370 | 73 | ||
| L10076.1 | - | 4 - 5 | 419 | 73 | ||
Set Membership: |
|