Search
Search

Ecel1| Gene Symbol: Ecel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277925.1 | NP_001264854.1 | 11 - 12 | 1955 | 65 | |
| NM_021306.3 | NP_067281.2 | 11 - 12 | 1947 | 65 | ||
| GenBank mRNA | AK039467.1 | - | 11 - 12 | 1948 | 65 | |
| AK159365.1 | - | 11 - 12 | 1957 | 65 | ||
| BC057569.1 | - | 10 - 11 | 1777 | 6413731 | 65 | |
Set Membership: |
|