Search
Search

Krt32| Gene Symbol: Krt32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159374.2 | NP_001152846.2 | 3 - 4 | 726 | 61 | |
| GenBank mRNA | BC117553.1 | - | 3 - 4 | 616 | 40104816 | 61 |
| BC118064.1 | - | 3 - 4 | 616 | 40104817 | 61 | |
| X75649.1 | - | 3 - 4 | 616 | 61 | ||
Set Membership: |
|