| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100034649.mm.1 |
NM_001102650.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100034649.mm.1 |
NM_001102650.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100034649.mm.1 |
NM_026561.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100034649.mm.1 |
NM_026561.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100034649.mm.1 |
TC1100003771.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100034651.mm.1 |
NM_001102650.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100034651.mm.1 |
NM_001102650.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100034651.mm.1 |
NM_026561.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100034651.mm.1 |
NM_026561.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100034651.mm.1 |
TC1100003771.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100034652.mm.1 |
NM_001102650.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100034652.mm.1 |
NM_001102650.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100034652.mm.1 |
NM_026561.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100034652.mm.1 |
NM_026561.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100034652.mm.1 |
TC1100003771.mm.1 |
MTA TC |