Search
Search

Fkbp10| Gene Symbol: Fkbp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163481.1 | NP_001156953.1 | 3 - 4 | 671 | 64 | |
| NM_010221.2 | NP_034351.2 | 3 - 4 | 671 | 64 | ||
| GenBank mRNA | AK019446.2 | - | 3 - 4 | 609 | 64 | |
| AK144185.1 | - | 3 - 4 | 654 | 64 | ||
| BC029546.1 | - | 3 - 4 | 618 | 5059670 | 64 | |
Set Membership: |
|