Search
Search
| Gene Symbol: Unkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197024.1 | NP_001183953.1 | 4 - 5 | 615 | 78 | |
| NM_001290736.1 | NP_001277665.1 | 4 - 5 | 626 | 78 | ||
| GenBank mRNA | BC059910.1 | - | 4 - 5 | 626 | 78 | |
Set Membership: |
|