Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01303078_m1
          See other ZFP207 GE Assays ›
          Gene Symbol:
          Zfp207
          Gene Name:
          zinc finger protein 207
          Gene Aliases:
          8430401D15Rik, BuGZ, Zep, Znf207
          Chromosome Location:
          Chr.11: 80383279 - 80405992 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01303078_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp207
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001130169.1 NP_001123641.1 3 - 4 483 106
          NM_001130170.1 NP_001123642.1 3 - 4 483 106
          NM_001130171.1 NP_001123643.1 3 - 4 483 106
          NM_011751.3 NP_035881.1 3 - 4 483 106
          NR_045038.1 3 - 4 483 106
          GenBank mRNA AB013357.1 - 3 - 4 417 106
          AB032196.1 - 3 - 4 364 106
          AK033497.1 - 3 - 4 432 106
          AK087885.1 - 3 - 4 483 106
          AK140139.1 - 3 - 4 466 107
          AK146787.1 - 3 - 4 446 106
          AK159683.1 - 3 - 4 434 106
          BB666896.1 - 3 - 4 483 106
          BC003715.1 - 3 - 4 424 3601357 106

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001204.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp207

          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic sister chromatid segregation
          cell cycle
          chromosome segregation
          mitotic nuclear division
          mitotic spindle assembly checkpoint
          attachment of spindle microtubules to kinetochore
          protein stabilization
          cell division
          regulation of chromosome segregation
          microtubule binding
          heparin binding
          poly(A) RNA binding
          metal ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100011260.mm.1 NM_001130169.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011260.mm.1 NM_001130169.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011260.mm.1 NM_001130170.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011260.mm.1 NM_001130170.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011260.mm.1 NM_001130171.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011260.mm.1 NM_001130171.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011260.mm.1 NM_011751.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011260.mm.1 NM_011751.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011260.mm.1 NR_045038.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011260.mm.1 NR_045038.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011260.mm.1 TC1100001204.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 376692 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline