Search
Search
| Gene Symbol: Lig3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291245.1 | NP_001278174.1 | 16 - 17 | 2510 | 97 | |
| NM_001291246.1 | NP_001278175.1 | 16 - 17 | 2507 | 97 | ||
| NM_001291247.1 | NP_001278176.1 | 16 - 17 | 2511 | 97 | ||
| NM_010716.3 | NP_034846.2 | 16 - 17 | 2514 | 97 | ||
| GenBank mRNA | AK031540.1 | - | 16 - 17 | 97 | ||
| AK143531.1 | - | 16 - 17 | 2380 | 97 | ||
| AK150644.1 | - | 16 - 17 | 2419 | 97 | ||
| AK156018.1 | - | 16 - 17 | 2429 | 97 | ||
| AK156030.1 | - | 5 - 6 | 496 | 97 | ||
| AK156825.1 | - | 5 - 6 | 496 | 97 | ||
| BC049240.1 | - | 16 - 17 | 2417 | 4504384 | 97 | |
| BC083500.1 | - | 16 - 17 | 2413 | 3673103 | 97 | |
| U66057.1 | - | 16 - 17 | 2448 | 97 | ||
| U66058.1 | - | 16 - 17 | 2448 | 97 | ||
Set Membership: |
|