Search
Search

Sept4| Gene Symbol: Sept4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284392.1 | NP_001271321.1 | 5 - 6 | 612 | 64 | |
| NM_001284394.1 | NP_001271323.1 | 6 - 7 | 909 | 64 | ||
| NM_001284398.1 | NP_001271327.1 | 5 - 6 | 596 | 64 | ||
| NM_011129.2 | NP_035259.1 | 6 - 7 | 909 | 64 | ||
| GenBank mRNA | AB078010.1 | - | 5 - 6 | 507 | 64 | |
| AK133933.1 | - | 5 - 6 | 564 | 64 | ||
| AK135143.1 | - | 6 - 7 | 910 | 64 | ||
| BC055101.1 | - | 6 - 7 | 909 | 6591016 | 64 | |
| X61452.1 | - | 6 - 7 | 839 | 64 | ||
Set Membership: |
|