Search
Search

Cacnb1| Gene Symbol: Cacnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159319.2 | NP_001152791.1 | 7 - 8 | 850 | 65 | |
| NM_145121.3 | NP_660099.2 | 7 - 8 | 850 | 65 | ||
| GenBank mRNA | AY094172.1 | - | 7 - 8 | 648 | 65 | |
| AY094173.1 | - | 7 - 8 | 648 | 65 | ||
| CA316737.1 | - | 6 - 7 | 544 | 65 | ||
Set Membership: |
|