Search
Search

Cacnb1| Gene Symbol: Cacnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282977.1 | NP_001269906.1 | 13 - 14 | 1666 | 99 | |
| NM_145121.3 | NP_660099.2 | 13 - 14 | 1531 | 99 | ||
| GenBank mRNA | AB685743.1 | - | 13 - 14 | 1650 | 99 | |
| AY094173.1 | - | 13 - 14 | 1329 | 99 | ||
Set Membership: |
|