Search
Search

Dhx40| Gene Symbol: Dhx40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026191.2 | NP_080467.3 | 15 - 16 | 2016 | 74 | |
| GenBank mRNA | AK004610.1 | - | 15 - 16 | 1980 | 74 | |
| AK010512.1 | - | 15 - 16 | 1996 | 74 | ||
| AK029935.1 | - | 15 - 16 | 1987 | 74 | ||
| AK031000.1 | - | 15 - 16 | 1962 | 74 | ||
| AK075991.1 | - | 9 - 10 | 1022 | 74 | ||
| AK135592.1 | - | 15 - 16 | 1953 | 74 | ||
| BC058276.1 | - | 15 - 16 | 1934 | 6515479 | 74 | |
Set Membership: |
|