Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01303999_m1
          See other 5430437J10RIK GE Assays ›
          Gene Symbol:
          5430437J10Rik
          Gene Name:
          RIKEN cDNA 5430437J10 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 5496318 - 5594983 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01303999_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 5430437J10Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045274.1 3 - 4 774 164
          GenBank mRNA AK017404.1 - 3 - 4 774 164

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500000025.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2389518 BC023070.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 829030 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 57632 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 97447 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 768836 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 225998 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 502570 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 858841 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 2982 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 726572 AK136532.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 726572 BC005434.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 726572 NM_009257.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 726572 U54705.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 785427 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 466437 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 884392 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 583410 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 806029 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 288356 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 885984 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 142776 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 345700 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 662400 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 449526 AJ293625.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 449526 AK028746.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 449526 BC048513.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 449526 BC065396.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 449526 BC100567.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 449526 BC116739.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 449526 BC116741.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 220961 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 859374 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 521250 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 220226 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 688827 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 364221 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 331895 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 539050 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 207122 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 699050 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 231819 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 37948 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 478509 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 525422 AK014899.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 690817 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 289203 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 452996 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 83852 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 38962 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 137991 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 129224 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 68045 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 169460 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 139763 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08271202 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08271102 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08271234 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08271154 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02964525 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02964479 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08271067 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101712 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08271102 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08271234 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08271154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12324300 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983922 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12088855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983810 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983989 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29984000 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12014571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12152128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983791 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12113440 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983971 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983803 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983796 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983808 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983784 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983982 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983778 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983789 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983931 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983977 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983781 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12446647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12298136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983788 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983783 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29984002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983987 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983973 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983976 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983929 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983813 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983797 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29983919 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1431577_PM_at - -
          Mouse Genome 430 2.0 Array 1431577_at - -
          Mouse Genome 430 2.0 Array 1431577_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1500000206.mm.1 NR_045274.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000206.mm.1 NR_045274.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000206.mm.1 TC1500000025.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500000202.mm.1 NR_045274.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000202.mm.1 NR_045274.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000202.mm.1 TC1500000025.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500000205.mm.1 NR_045274.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000205.mm.1 NR_045274.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000205.mm.1 TC1500000025.mm.1 MTA TC
          Mouse Expression Set 430 1431577_PM_at - -
          Mouse Expression Set 430 1431577_at - -
          Affymetrix HT Mouse Genome 430B Array 1431577_at - -
          Affymetrix HT Mouse Genome 430B Array 1431577_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 679535 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline