Search
Search

Habp4| Gene Symbol: Habp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019986.3 | NP_064370.2 | 5 - 6 | 942 | 92 | |
| GenBank mRNA | AF227684.1 | - | 5 - 6 | 885 | 92 | |
| AK016800.1 | - | 5 - 6 | 985 | 92 | ||
| AK144206.1 | - | 5 - 6 | 926 | 92 | ||
| AK146525.1 | - | 5 - 6 | 915 | 92 | ||
| BC082806.1 | - | 5 - 6 | 944 | 92 | ||
| BC148304.2 | - | 4 - 5 | 446 | 92 | ||
Set Membership: |
|