Search
Search
| Gene Symbol: Dlst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030225.4 | NP_084501.1 | 2 - 3 | 160 | 94 | |
| GenBank mRNA | AK019713.2 | - | 157 | 94 | ||
| AK054053.1 | - | 2 - 3 | 114 | 94 | ||
| AK149664.1 | - | 2 - 3 | 136 | 94 | ||
| AK158877.1 | - | 2 - 3 | 161 | 94 | ||
| AK168570.1 | - | 2 - 3 | 136 | 94 | ||
| AK169943.1 | - | 2 - 3 | 114 | 94 | ||
| BC006702.1 | - | 2 - 3 | 97 | 3583936 | 94 | |
| CT010197.1 | - | 2 - 3 | 96 | 94 | ||
Set Membership: |
|