Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01304910_m1
          See other B230217C12RIK GE Assays ›
          Gene Symbol:
          B230217C12Rik
          Gene Name:
          RIKEN cDNA B230217C12 gene
          Gene Aliases:
          AI840637
          Chromosome Location:
          Chr.11: 97840780 - 97843043 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01304910_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price 434.00
          Your Price
          Online offer: 407.65
          407.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: B230217C12Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK018341.1 - 1 - 2 252 84
          AK031146.1 - 251 84
          AK042827.1 - 254 84
          AK080808.1 - 251 84
          AK081899.1 - 272 84
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY726757.1 - 1 - 2 224 Chr. 11 - 97841341 - 97842619 [+] on Build GRCm38 84
          GenBankmRNA BY123368.1 - 1 - 2 250 Chr. 11 - 97841315 - 97842277 [+] on Build GRCm38 84
          GenBankmRNA BE864068.1 - 1 - 2 235 Chr. 11 - 97841330 - 97842314 [+] on Build GRCm38 84
          GenBankmRNA AU079083.1 - Chr. - - [-] on Build 84
          GenBankmRNA BY251962.1 - 1 - 2 281 Chr. 11 - 97841285 - 97842344 [+] on Build GRCm38 84
          GenBankmRNA BB644141.1 - 1 - 2 251 Chr. 11 - 97841316 - 97842576 [+] on Build GRCm38 84
          GenBankmRNA BY724878.1 - 1 - 2 250 Chr. 11 - 97841315 - 97842603 [+] on Build GRCm38 84
          GenBankmRNA BY774520.1 - 1 - 2 250 Chr. 11 - 97841315 - 97842298 [+] on Build GRCm38 84
          GenBankmRNA BE859964.1 - 1 - 2 235 Chr. 11 - 97841330 - 97842441 [+] on Build GRCm38 84
          GenBankmRNA BB620407.1 - 1 - 2 251 Chr. 11 - 97841315 - 97842838 [+] on Build GRCm38 84
          GenBankmRNA BB648198.1 - 1 - 2 272 Chr. 11 - 97841293 - 97842578 [+] on Build GRCm38 84
          GenBankmRNA BY719251.1 - 1 - 2 252 Chr. 11 - 97841316 - 97842774 [+] on Build GRCm38 84

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream LIM and SH3 protein 1 Lasp1 AA408629,
          Def-4,
          SH3P6,
          Tg(Col1a1-lacZ)1Ngma
          -2.6kb Sense Chr. 11 - 97799005 - 97838765 on NCBI Build GRCm38
          Downstream F-box protein 47 Fbxo47 2900052P03Rik,
          Gm1562
          8.7kb Anti-sense Chr. 11 - 97851558 - 97884390 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          ABI

          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 913925 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline