Search
Search

Tom1l1| Gene Symbol: Tom1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028011.2 | NP_082287.1 | 10 - 11 | 1079 | 113 | |
| GenBank mRNA | AF395837.1 | - | 10 - 11 | 1043 | 113 | |
| AJ505007.1 | - | 10 - 11 | 1040 | 113 | ||
| AK009826.1 | - | 8 - 9 | 814 | 113 | ||
| AK033712.1 | - | 10 - 11 | 1081 | 113 | ||
| AK083630.1 | - | 1041 | 113 | |||
| AK197727.1 | - | 2 - 3 | 137 | 113 | ||
| BC004710.1 | - | 3 - 4 | 1436 | 3499918 | 113 | |
Set Membership: |
|