Search
Search

Atp6v1d| Gene Symbol: Atp6v1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023721.2 | NP_076210.1 | 6 - 7 | 635 | 92 | |
| GenBank mRNA | AF298810.1 | - | 6 - 7 | 456 | 92 | |
| AK003446.1 | - | 3 - 4 | 172 | 92 | ||
| AK146129.1 | - | 4 - 5 | 306 | 92 | ||
| AK146448.1 | - | 6 - 7 | 538 | 92 | ||
| AK147910.1 | - | 552 | 92 | |||
| AK150981.1 | - | 6 - 7 | 524 | 92 | ||
| AK152367.1 | - | 6 - 7 | 524 | 92 | ||
| AK153250.1 | - | 6 - 7 | 524 | 92 | ||
| AK165761.1 | - | 6 - 7 | 529 | 92 | ||
| BC033457.1 | - | 6 - 7 | 497 | 3991107 | 92 | |
Set Membership: |
|