Search
Search

Rnft1| Gene Symbol: Rnft1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029788.5 | NP_084064.1 | 7 - 8 | 1148 | 138 | |
| GenBank mRNA | AK002624.1 | - | 7 - 8 | 1106 | 138 | |
| AK036401.1 | - | 7 - 8 | 1149 | 138 | ||
| AK153293.1 | - | 7 - 8 | 1117 | 138 | ||
| AK171077.1 | - | 6 - 7 | 2399 | 138 | ||
| BC037057.1 | - | 6 - 7 | 935 | 138 | ||
| BC050796.1 | - | 7 - 8 | 1116 | 6743413 | 138 | |
Set Membership: |
|