Search
Search
| Gene Symbol: Fmo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008030.1 | NP_032056.1 | 5 - 6 | 664 | 66 | |
| GenBank mRNA | AK050123.1 | - | 5 - 6 | 693 | 66 | |
| AK149440.1 | - | 5 - 6 | 691 | 66 | ||
| U87147.1 | - | 5 - 6 | 664 | 66 | ||
Set Membership: |
|