Search
Search

Zfyve1| Gene Symbol: Zfyve1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183154.3 | NP_898977.2 | 6 - 7 | 2009 | 61 | |
| GenBank mRNA | AK090334.1 | - | 7 - 8 | 2180 | 61 | |
| AK129400.1 | - | 6 - 7 | 1984 | 61 | ||
| AK139825.1 | - | 6 - 7 | 2030 | 61 | ||
| AK170954.1 | - | 6 - 7 | 2010 | 61 | ||
| BC050015.1 | - | 5 - 6 | 1430 | 30016571 | 61 | |
Set Membership: |
|