| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500000270.mm.1 |
NM_013485.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000270.mm.1 |
NM_013485.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000270.mm.1 |
TC1500000034.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500000269.mm.1 |
NM_013485.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000269.mm.1 |
NM_013485.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000269.mm.1 |
TC1500000034.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500000272.mm.1 |
NM_013485.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000272.mm.1 |
NM_013485.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500000272.mm.1 |
TC1500000034.mm.1 |
MTA TC |