Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01307530_s1
          See other FAM58B GE Assays ›
          Gene Symbol:
          Fam58b
          Gene Name:
          family with sequence similarity 58, member B
          Gene Aliases:
          1810009O10Rik, Fam58a
          Chromosome Location:
          Chr.11: 78750506 - 78751729 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01307530_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam58b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_197989.1 NP_932106.1 1 879 72
          GenBank mRNA AK032408.1 - 834 72
          BC027022.1 - 1 879 5367363 72

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fam58b

          Panther Classification:

          Molecular Function -

          kinase activator

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cyclin-dependent protein serine/threonine kinase activity
          regulation of transcription, DNA-templated
          positive regulation of MAPK cascade
          positive regulation of protein kinase activity
          protein binding
          protein kinase binding
          protein kinase activator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27945142 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27945140 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27945141 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2013598 AK032408.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2013598 BC027022.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2013598 NM_197989.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2013598 AK007413.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 336768 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2013598 AK032408.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2013598 BC027022.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2013598 NM_197989.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2013598 AK007413.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27945145 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27945146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27945128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27945125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27945144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27945147 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27945156 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27945126 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27945141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27945129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27945151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27945149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27945142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27945158 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27945150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27945152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27945130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27945154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27945160 - -
          Whole Mouse Genome G4122A A_52_P595619 AK032408.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P595619 BC027022.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P595619 NM_197989.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P595619 AK007413.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09459738 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09166313 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09166322 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09166326 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09459735 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27945142 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27945141 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27945140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27945175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09160092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09524770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09706607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09524775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09348161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09034401 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09706611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09670623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09328154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09328158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09166324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09557253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09524769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09328740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09316532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09498863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27945126 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10388899 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388901 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388900 - -
          Affymetrix HT MG-430 PM Array Plate 1451334_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10388898 - -
          Mouse Genome 430 2.0 Array 1451334_at - -
          Mouse Genome 430 2.0 Array 1451334_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69109_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69109_at - -
          Mouse Expression Set 430 1451334_at - -
          Mouse Expression Set 430 1451334_PM_at - -
          Murine Genome U74v2 Set 168538_r_at - -
          Murine Genome U74v2 Set 168538_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1451334_PM_at - -
          Mouse Genome 430A 2.0 Array 1451334_at - -
          Affymetrix HT Mouse Genome 430A Array 1451334_at - -
          Affymetrix HT Mouse Genome 430A Array 1451334_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 894397 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4570368 - -
          Illumina mouse-6 v1.0 expression beadchip 101050022 - -
          Illumina mouse-6 v1.0 expression beadchip 4570368 - -
          Illumina mouse-6 v1.0 expression beadchip 360324 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004570368 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000360324 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2742294 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2757087 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2742294 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2757087 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2498219 - -
          mouse-6 v1.1 (Illumina) scl0069109.2_3-S - -
          mouse-6 v1.1 (Illumina) scl0069109.2_11-S - -
          mouse-6 v1.1 (Illumina) scl39874.1_89-S - -
          Illumina Sentrix mouse-6.v1 bead array 360324 - -
          Illumina Sentrix mouse-6.v1 bead array 101050022 - -
          Illumina Sentrix mouse-6.v1 bead array 4570368 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0069109.2_11-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0069109.2_3-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4570368 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 360324 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101050022 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2742293 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2757087 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2498219 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2742293 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2757087 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0069109.2_11-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline