Search
Search

Mmp25| Gene Symbol: Mmp25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033339.3 | NP_001028511.1 | 1 - 2 | 678 | 72 | |
| GenBank mRNA | AK089273.1 | - | 1 - 2 | 418 | 72 | |
| AK089533.1 | - | 1 - 2 | 418 | 72 | ||
| AK137491.1 | - | 1 - 2 | 386 | 72 | ||
| AK154458.1 | - | 1 - 2 | 680 | 72 | ||
| AK171493.1 | - | 1 - 2 | 418 | 72 | ||
| BC059059.1 | - | 1 - 2 | 652 | 72 | ||
| BC112379.1 | - | 1 - 2 | 364 | 40059601 | 72 | |
Set Membership: |
|