Search
Search

Mmp25| Gene Symbol: Mmp25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033339.3 | NP_001028511.1 | 4 - 5 | 1235 | 91 | |
| GenBank mRNA | AK089273.1 | - | 4 - 5 | 974 | 91 | |
| AK089533.1 | - | 4 - 5 | 974 | 91 | ||
| AK137491.1 | - | 4 - 5 | 837 | 91 | ||
| AK154458.1 | - | 4 - 5 | 1237 | 91 | ||
| AK171493.1 | - | 4 - 5 | 974 | 91 | ||
| BC059059.1 | - | 3 - 4 | 1305 | 91 | ||
| BC112379.1 | - | 4 - 5 | 921 | 40059601 | 91 | |
Set Membership: |
|