Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01309402_m1
          See other TMSB15A GE Assays ›
          Gene Symbol:
          Tmsb15a
          Gene Name:
          thymosin beta 15a
          Gene Aliases:
          1700129I15Rik, Tb15a, Tmsbl1, Tmsl8
          Chromosome Location:
          Chr.X: 135718667 - 135725261 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01309402_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmsb15a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030106.2 NP_084382.1 2 - 3 214 124
          GenBank mRNA AK018998.1 - 2 - 3 216 124
          AK028086.1 - 2 - 3 178 124

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00003015.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmsb15a

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2185689 AK018998.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2185689 AK028086.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2185689 NM_030106.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 432397 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 117179 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P112359 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2185689 AK018998.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2185689 AK028086.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2185689 NM_030106.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677990 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677976 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677995 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677961 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677970 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677989 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677967 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677996 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677969 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677971 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32678001 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677998 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677964 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677984 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677988 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677993 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677963 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677962 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P112359 - -
          Whole Mouse Genome G4122A A_51_P194597 AK018998.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P194597 AK028086.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P194597 NM_030106.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134727 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119367 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P112359 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14564400 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14564395 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14564401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32678025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32678023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32678004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32678029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32678035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32678027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32678007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32678012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32678020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32678010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32678009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32678026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32678030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32678001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32678033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32678032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32678031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32678011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15088669 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677982 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32678035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677973 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14720477 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14641683 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14564397 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14635692 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32678007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14564401 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677960 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15107651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14564405 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32678027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32678009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32678030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14959675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32678029 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32678033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32678023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32678032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32678031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677957 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677998 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14635691 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32678020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32678004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677989 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15088674 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677977 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32678001 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10606833 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606832 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606834 - -
          Affymetrix HT MG-430 PM Array Plate 1432566_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10606831 - -
          Mouse Genome 430 2.0 Array 1432566_at - -
          Mouse Genome 430 2.0 Array 1432566_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78478_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78478_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0X00018892.mm.1 NM_030106.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00018892.mm.1 NM_030106.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00018892.mm.1 TC0X00003015.mm.1 MTA TC
          Mouse Expression Set 430 1432566_PM_at - -
          Mouse Expression Set 430 1432566_at - -
          Affymetrix HT Mouse Genome 430B Array 1432566_at - -
          Affymetrix HT Mouse Genome 430B Array 1432566_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 821399 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1252841 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2793688 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2793688 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline