Search
Search

Utp18| Gene Symbol: Utp18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013375.1 | NP_001013393.1 | 9 - 10 | 1308 | 95 | |
| GenBank mRNA | AK032265.1 | - | 8 - 9 | 1835 | 95 | |
| AK078033.1 | - | 4 - 5 | 444 | 95 | ||
| AK154637.1 | - | 9 - 10 | 1248 | 95 | ||
| AK167248.1 | - | 9 - 10 | 1307 | 95 | ||
| BC119621.1 | - | 9 - 10 | 1281 | 40062238 | 95 | |
| BC119622.1 | - | 9 - 10 | 1281 | 40062245 | 95 | |
Set Membership: |
|