Search
Search

Slc35f5| Gene Symbol: Slc35f5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028787.4 | NP_083063.2 | 14 - 15 | 1725 | 90 | |
| GenBank mRNA | AK004892.2 | - | 14 - 15 | 1706 | 90 | |
| AK076339.1 | - | 14 - 15 | 1720 | 90 | ||
| AK145848.1 | - | 14 - 15 | 1726 | 90 | ||
| AK169012.1 | - | 14 - 15 | 1587 | 90 | ||
| AK171951.1 | - | 14 - 15 | 1699 | 90 | ||
| BC026858.1 | - | 14 - 15 | 1584 | 4014064 | 90 | |
Set Membership: |
|