Search
Search

Stxbp4| Gene Symbol: Stxbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011505.2 | NP_035635.1 | 6 - 7 | 727 | 94 | |
| GenBank mRNA | AF152924.1 | - | 6 - 7 | 717 | 94 | |
| AK049001.1 | - | 6 - 7 | 696 | 94 | ||
| BC032881.1 | - | 6 - 7 | 681 | 1224878 | 94 | |
Set Membership: |
|