Search
Search

Lrrc63| Gene Symbol: Lrrc63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027581.1 | NP_081857.1 | 3 - 4 | 985 | 181 | |
| GenBank mRNA | AK014850.1 | - | 2 - 3 | 717 | 181 | |
| BC145796.1 | - | 3 - 4 | 40131099 | 178 | ||
Set Membership: |
|