Search
Search

Ero1l| Gene Symbol: Ero1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015774.3 | NP_056589.1 | 5 - 6 | 513 | 63 | |
| GenBank mRNA | AK142595.1 | - | 5 - 6 | 504 | 63 | |
| AK159217.1 | - | 5 - 6 | 480 | 63 | ||
| BC025102.1 | - | 5 - 6 | 465 | 2654098 | 63 | |
| CT010195.1 | - | 5 - 6 | 431 | 63 | ||
| CT010259.1 | - | 5 - 6 | 431 | 63 | ||
Set Membership: |
|