Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01311042_g1
          See other 1810026B05RIK GE Assays ›
          Gene Symbol:
          1810026B05Rik
          Gene Name:
          RIKEN cDNA 1810026B05 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 73539798 - 73558395 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01311042_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1810026B05Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_037569.1 2 - 3 426 134
          GenBank mRNA AK140938.1 - 2 - 3 426 134
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AI854867.1 - 2 - 3 279 Chr. 7 - 73549600 - 73558247 [-] on Build GRCm38 134
          GenBankmRNA BE691918.1 - 2 - 3 232 Chr. 7 - 73549612 - 73558201 [-] on Build GRCm38 134
          GenBankmRNA CF198176.1 - 1 - 2 82 Chr. 7 - 73544037 - 73555921 [-] on Build GRCm38 134
          GenBankmRNA CR516393.1 - 3 - 4 Chr. 7 - 73544304 - 73558084 [+] on Build GRCm38 134
          GenBankmRNA BE457530.1 - 2 - 3 271 Chr. 7 - 73549608 - 73558240 [-] on Build GRCm38 134
          GenBankmRNA BE685660.1 - 2 - 3 280 Chr. 7 - 73549624 - 73558249 [-] on Build GRCm38 134
          GenBankmRNA CK791988.1 - 3 - 4 Chr. 7 - 73544036 - 73558178 [+] on Build GRCm38 134
          GenBankmRNA BG276399.1 - 3 - 4 Chr. 7 - 73544140 - 73558077 [+] on Build GRCm38 134
          GenBankmRNA CB248644.1 - 2 - 3 372 Chr. 7 - 73544231 - 73558341 [-] on Build GRCm38 134
          GenBankmRNA BG093603.1 - 2 - 3 280 Chr. 7 - 73544250 - 73558249 [-] on Build GRCm38 134
          GenBankmRNA AW495687.1 - 2 - 3 279 Chr. 7 - 73544314 - 73558248 [-] on Build GRCm38 134
          GenBankmRNA BB649475.1 - 2 - 3 422 Chr. 7 - 73544318 - 73558391 [-] on Build GRCm38 134
          GenBankmRNA BE630945.1 - 2 - 3 243 Chr. 7 - 73544320 - 73558212 [-] on Build GRCm38 134
          GenBankmRNA CJ236647.1 - 2 - 3 137 Chr. 7 - 73544036 - 73558098 [-] on Build GRCm38 134
          GenBankmRNA AI006392.1 - 3 - 4 254 Chr. 7 - 73555249 - 73558369 [-] on Build GRCm38 134
          GenBankmRNA BG089011.1 - 3 - 4 Chr. 7 - 73544192 - 73558101 [+] on Build GRCm38 134
          GenBankmRNA BP427421.1 - 2 - 3 134 Chr. 7 - 73555152 - 73558100 [-] on Build GRCm38 134
          GenBankmRNA AI550450.1 - 2 - 3 280 Chr. 7 - 73544296 - 73558249 [-] on Build GRCm38 134
          GenBankmRNA BY098640.1 - 2 - 3 136 Chr. 7 - 73544299 - 73558104 [-] on Build GRCm38 134
          GenBankmRNA BE685646.1 - 2 - 3 280 Chr. 7 - 73555255 - 73558249 [-] on Build GRCm38 134
          GenBankmRNA BE985864.1 - 2 - 3 279 Chr. 7 - 73544122 - 73558248 [-] on Build GRCm38 134
          GenBankmRNA AA214922.1 - 2 - 3 Chr. 7 - 73549597 - 73558202 [+] on Build GRCm38 134
          GenBankmRNA BE627877.1 - 2 - 3 280 Chr. 7 - 73544310 - 73558249 [-] on Build GRCm38 134

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream chromodomain helicase DNA binding protein 2 Chd2 2810013C04Rik,
          2810040A01Rik,
          5630401D06Rik,
          AI851092,
          BC029703,
          CHD-2
          -13.5kb Anti-sense Chr. 7 - 73426652 - 73541746 on NCBI Build GRCm38
          Downstream Smad nuclear interacting protein 1 pseudogene Gm4971 EG244061 129.6kb Anti-sense Chr. 7 - 73685492 - 73686563 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700003493.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700029707.mm.1 NR_037569.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700029707.mm.1 NR_037569.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700029707.mm.1 TC0700003493.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline