Search
Search

Samd4| Gene Symbol: Samd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037221.2 | NP_001032298.1 | 11 - 12 | 2645 | 63 | |
| NM_001163433.1 | NP_001156905.1 | 10 - 11 | 2125 | 63 | ||
| NM_028966.3 | NP_083242.1 | 10 - 11 | 2381 | 63 | ||
| GenBank mRNA | AK017081.1 | - | 6 - 7 | 959 | 63 | |
| AK034323.1 | - | 9 - 10 | 1975 | 63 | ||
| AK133470.1 | - | 10 - 11 | 2361 | 63 | ||
| AK134243.1 | - | 11 - 12 | 2643 | 63 | ||
| AK161277.1 | - | 10 - 11 | 2119 | 63 | ||
| BC049740.1 | - | 6 - 7 | 931 | 63 | ||
| BC145827.1 | - | 10 - 11 | 2247 | 40131130 | 63 | |
| BC145829.1 | - | 10 - 11 | 2246 | 40131132 | 63 | |
| DQ278487.1 | - | 10 - 11 | 2158 | 63 | ||
Set Membership: |
|