Search
Search

Samd4| Gene Symbol: Samd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037221.2 | NP_001032298.1 | 7 - 8 | 2121 | 131 | |
| NM_001163433.1 | NP_001156905.1 | 6 - 7 | 1601 | 131 | ||
| NM_028966.3 | NP_083242.1 | 6 - 7 | 1857 | 131 | ||
| GenBank mRNA | AK017081.1 | - | 2 - 3 | 435 | 131 | |
| AK034323.1 | - | 5 - 6 | 1451 | 131 | ||
| AK133470.1 | - | 6 - 7 | 1836 | 131 | ||
| AK134243.1 | - | 7 - 8 | 2119 | 131 | ||
| AK161277.1 | - | 6 - 7 | 1595 | 131 | ||
| BC049740.1 | - | 2 - 3 | 407 | 131 | ||
| BC145827.1 | - | 6 - 7 | 1723 | 40131130 | 131 | |
| BC145829.1 | - | 6 - 7 | 1722 | 40131132 | 131 | |
| DQ278487.1 | - | 6 - 7 | 1634 | 131 | ||
Set Membership: |
|