Search
Search

Ap3b2| Gene Symbol: Ap3b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021492.3 | NP_067467.2 | 14 - 15 | 1972 | 66 | |
| GenBank mRNA | AK134504.1 | - | 14 - 15 | 1721 | 66 | |
| AY528675.1 | - | 14 - 15 | 1753 | 66 | ||
| BC139378.1 | - | 14 - 15 | 1720 | 8862400 | 66 | |
| BC139379.1 | - | 14 - 15 | 1720 | 8862401 | 66 | |
Set Membership: |
|