Search
Search

Plxna2| Gene Symbol: Plxna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008882.2 | NP_032908.2 | 14 - 15 | 3498 | 60 | |
| GenBank mRNA | AK122289.1 | - | 13 - 14 | 1762 | 60 | |
| BC068155.1 | - | 14 - 15 | 3280 | 30362726 | 60 | |
| D86949.1 | - | 14 - 15 | 3404 | 60 | ||
Set Membership: |
|