Search
Search

Slc35a3| Gene Symbol: Slc35a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144902.3 | NP_659151.1 | 6 - 7 | 967 | 114 | |
| GenBank mRNA | AK029398.1 | - | 6 - 7 | 1339 | 114 | |
| AK041195.1 | - | 7 - 8 | 1087 | 114 | ||
| AK049484.1 | - | 6 - 7 | 967 | 114 | ||
| AK161317.1 | - | 6 - 7 | 925 | 114 | ||
| BC024110.1 | - | 6 - 7 | 963 | 5150304 | 114 | |
Set Membership: |
|