Search
Search
| Gene Symbol: Brox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027861.2 | NP_082137.1 | 10 - 11 | 968 | 90 | |
| GenBank mRNA | AK036197.1 | - | 4 - 5 | 303 | 90 | |
| AK050525.1 | - | 10 - 11 | 939 | 90 | ||
| AK146863.1 | - | 9 - 10 | 828 | 90 | ||
| AK162247.1 | - | 10 - 11 | 938 | 90 | ||
| BC030340.1 | - | 10 - 11 | 974 | 3498026 | 90 | |
| BC120692.1 | - | 9 - 10 | 816 | 40129615 | 90 | |
| BC125527.1 | - | 9 - 10 | 816 | 40130042 | 90 | |
Set Membership: |
|