| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1400018692.mm.1 |
NM_001033439.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018692.mm.1 |
NM_001033439.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018692.mm.1 |
NM_001252132.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018692.mm.1 |
NM_001252132.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018692.mm.1 |
TC1400002406.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400018691.mm.1 |
NM_001033439.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018691.mm.1 |
NM_001033439.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018691.mm.1 |
NM_001252132.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018691.mm.1 |
NM_001252132.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018691.mm.1 |
TC1400002406.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400018690.mm.1 |
NM_001033439.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018690.mm.1 |
NM_001033439.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018690.mm.1 |
TC1400002406.mm.1 |
MTA TC |