Search
Search

Mcoln3| Gene Symbol: Mcoln3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134160.1 | NP_598921.1 | 3 - 4 | 546 | 67 | |
| GenBank mRNA | AF475086.1 | - | 3 - 4 | 546 | 67 | |
| AK033008.1 | - | 3 - 4 | 660 | 67 | ||
| AK035029.1 | - | 3 - 4 | 667 | 67 | ||
| AY083531.1 | - | 3 - 4 | 544 | 67 | ||
| BC106856.2 | - | 3 - 4 | 546 | 40047039 | 67 | |
| BC106857.2 | - | 3 - 4 | 546 | 40047042 | 67 | |
Set Membership: |
|