| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300023882.mm.1 |
NM_001040690.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300023882.mm.1 |
NM_001286759.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300023882.mm.1 |
NM_145544.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300023882.mm.1 |
TC0300003023.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300023880.mm.1 |
NM_001040690.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300023880.mm.1 |
NM_001286759.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300023880.mm.1 |
NM_145544.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300023880.mm.1 |
TC0300003023.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300023878.mm.1 |
NM_001040690.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300023878.mm.1 |
NM_001286759.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300023878.mm.1 |
NM_145544.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300023878.mm.1 |
TC0300003023.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300023881.mm.1 |
NM_001040690.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300023881.mm.1 |
NM_001286759.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300023881.mm.1 |
NM_145544.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300023881.mm.1 |
TC0300003023.mm.1 |
MTA TC |