Search
Search

Rap1gds1| Gene Symbol: Rap1gds1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040690.2 | NP_001035780.1 | 8 - 9 | 1066 | 80 | |
| NM_001286759.1 | NP_001273688.1 | 8 - 9 | 1066 | 80 | ||
| NM_145544.3 | NP_663519.2 | 7 - 8 | 919 | 80 | ||
| GenBank mRNA | AK052125.1 | - | 7 - 8 | 867 | 80 | |
| AK154852.1 | - | 8 - 9 | 1052 | 80 | ||
| AK155793.1 | - | 8 - 9 | 1052 | 80 | ||
| AK160989.1 | - | 8 - 9 | 1002 | 80 | ||
| AK164159.1 | - | 8 - 9 | 978 | 80 | ||
| AK166315.1 | - | 8 - 9 | 999 | 80 | ||
| AK172052.1 | - | 8 - 9 | 1068 | 80 | ||
| AK197097.1 | - | 1 - 2 | 50 | 80 | ||
| BC011279.1 | - | 7 - 8 | 788 | 4008751 | 80 | |
Set Membership: |
|